Ethyl-4-bromobenzoate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromobenzoic acid ethyl ester, Ethyl 4-bromobenzoate

24976-0500 24976-0100
TS24976-0500EA 132 USD
TS24976-0500 TS24976-0100
Ethyl-4-bromobenzoate 99%
Ethyl-4-bromobenzoate
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 262…264 °C
Melting Pt: 18 °C
Density: 1.43 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00016329
CAS Number: 5798-75-4
EINECS: 227-347-2
Merck Index: 13,01392

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5440 to 1.5470 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR