(±)-1-Methoxy-2-propanol 98.5%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

24499-0010 24499-0025 24499-0050 24499-0250 24499-0251
TS24499-0010EA 61 USD
TS24499-0010 TS24499-0025 TS24499-0050 TS24499-0250 TS24499-0251
(±)-1-Methoxy-2-propanol 98.5%, Extra Pure
(±)-1-Methoxy-2-propanol
Formula: CH₃CH(OH)CH₂OCH₃
Boiling Pt: 120 °C (760 mmHg)
Melting Pt: –97 °C
Density: 0.92 g/cm³
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00004537
CAS Number: 107-98-2
EINECS: 203-539-1
UN: 3092
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9e076d8e7a4ef95472e37d2c355f37d64eac8ee&vid=26da88ae9f09b8195abc9bdd949f6bbdaf429255&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4020 to 1.4040 (20°C, 589 nm)
Water ≤0.1 %
GC ≥98.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.01 %
Acidity (CH3COOH) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR