Methyl-3,4,5-trihydroxybenzoate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl gallate, Gallic acid methyl ester, Methyl 3,4,5-trihydroxybenzoate

24346-0500 24346-0050 24346-2500
TS24346-0500EA 43 USD
TS24346-0500 TS24346-0050 TS24346-2500
Methyl-3,4,5-trihydroxybenzoate 99%
Methyl-3,4,5-trihydroxybenzoate
Formula: C₈H₈O₅
Melting Pt: 199…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00002194
CAS Number: 99-24-1
EINECS: 202-741-7
Merck Index: 13,04366

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 199 to 203 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR