tert-Butyl-4-methoxyphenol (mixture of isomers) 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-Butyl-4-methoxyphenol, BHA, tert-Butyl-4-hydroxyanisole, Butylhydroxyanisole, 2(3)-tert-Butyl-4-methoxyphenol, Butylated hydroxyanisole

23523-1000 23523-0025 23523-5000
TS23523-1000EA 39 USD
TS23523-1000 TS23523-0025 TS23523-5000
tert-Butyl-4-methoxyphenol (mixture of isomers) 96%
tert-Butyl-4-methoxyphenol (mixture of isomers)
Formula: C₁₁H₁₆O₂
Boiling Pt: 268 °C (760 mmHg)
Melting Pt: 48…55 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD01779059
CAS Number: 25013-16-5
EINECS: 246-563-8
Merck Index: 12,01582

Order Now

Specification Test Results

Appearance White to light yellow Crystals or flakes
Infrared spectrum Conforms
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR