O-Phenetidine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Ethoxyaniline, 2-Aminophenetole

23348-0050 23348-5000
TS23348-0050EA 21 USD
TS23348-0050 TS23348-5000
O-Phenetidine 99%
O-Phenetidine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 229 °C (1013 hPa)
Melting Pt: –20 °C
Density: 1.05 g/cm³
Flash Pt: 80 °C
MDL Number: MFCD00007689
CAS Number: 94-70-2
EINECS: 202-356-4
UN: 2311
ADR: 6.1,III
Merck Index: 12,07372

Order Now

Specification Test Results

Appearance Light yellow to red Liquid
Infrared spectrum Conforms
Refractive index 1.5540 to 1.5560 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR