(S)-(-)-1,1'-Bi-2-naphthol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-BINOL, (S)-(-)-1,1'-Binaphthyl-2,2'-Diol, (S)-[1,1'-Binaphthalene]-2,2'-diol

22431-0050 22431-0010 22431-0250
TS22431-0050EA 130 USD
TS22431-0050 TS22431-0010 TS22431-0250
(S)-(-)-1,1'-Bi-2-naphthol 99%
(S)-(-)-1,1'-Bi-2-naphthol
Formula: C₂₀H₁₄O₂
Melting Pt: 205…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00004068
CAS Number: 18531-99-2

Order Now

Specification Test Results

Appearance White to light grey Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 205 to 211 °C
HPLC ≥98.5 %
Specific optical rotation -33° to -38° (20 °C, 589 nm) (c=1, THF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR