3-Methoxyphenyl isocyanate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Isocyanato-3-methoxybenzene

22116-0050 22116-0250
TS22116-0050EA 53 USD
TS22116-0050 TS22116-0250
3-Methoxyphenyl isocyanate 99%
3-Methoxyphenyl isocyanate
Formula: C₈H₇NO₂
MW: 149.15 g/mol
Boiling Pt: 94…95 °C (10 mmHg)
Density: 1.13 g/cm³
Flash Pt: 96 °C
MDL Number: MFCD00002019
CAS Number: 18908-07-1
EINECS: 242-658-3
UN: 2206
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to light pink Liquid
Infrared spectrum Conforms
Refractive index 1.5420 to 1.5440 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR