Gallic acid monohydrate

Supplier: Thermo Scientific Chemicals
Warning

22076-1000 22076-0025 22076-5000
TS22076-1000EA 103 USD
TS22076-1000 TS22076-0025 TS22076-5000
Gallic acid monohydrate
Formula: C₇H₆O₅·1H₂O
Melting Pt: 251 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149098
CAS Number: 5995-86-8
EINECS: 205-749-9

Order Now

Specification Test Results

Appearance Off-white to beige Powder
Infrared spectrum Conforms
Loss on drying ≤10 % (120°C)
Sulfated ash ≤0.1 %
Titration with NaOH ≥99 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR