4-Methoxyphenyl isocyanate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Isocyanato-4-methoxybenzene

21876-0050 21876-0250
TS21876-0050EA 56 USD
TS21876-0050 TS21876-0250
4-Methoxyphenyl isocyanate 99%
4-Methoxyphenyl isocyanate
Formula: C₈H₇NO₂
MW: 149.15 g/mol
Boiling Pt: 106…110 °C (16 mmHg)
Density: 1.15 g/cm³
Flash Pt: 98 °C
MDL Number: MFCD00002026
CAS Number: 5416-93-3
EINECS: 226-513-1
UN: 2206
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5470 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR