(2R,3R)-(-)-2,3-Butanediol 98+%, 99+% ee

Supplier: Thermo Scientific Chemicals

21798-0010 21798-0050
TS21798-0010EA 67 USD
TS21798-0010 TS21798-0050
(2R,3R)-(-)-2,3-Butanediol 98+%, 99+% ee
(2R,3R)-(-)-2,3-Butanediol
Formula: C₄H₁₀O₂
Boiling Pt: 77.3…77.4 °C (10 mmHg)
Melting Pt: 16 °C
Density: 0.98 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00064267
CAS Number: 24347-58-8
EINECS: 246-186-9
Merck Index: 13,01567

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4309 to 1.4329 (20°C, 589 nm)
GC ≥98.0 %
Enantiomeric excess ≥99.0 % (GC)
Specific optical rotation -12.5° to -13.5° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR