1-Phenyl-1,2-propanedione 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Phenylpropane-1,2-dione

20736-0010 20736-0050
TS20736-0010EA 36 USD
TS20736-0010 TS20736-0050
1-Phenyl-1,2-propanedione 98%
1-Phenyl-1,2-propanedione
Formula: C₆H₅C(O)C(O)CH₃
MW: 148.16 g/mol
Boiling Pt: 103…105 °C (14 mmHg)
Density: 1.1 g/cm³
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00008755
CAS Number: 579-07-7
EINECS: 209-435-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1394d5392b739ebd2496078378d299949ea575ae&vid=b85d2a588e386bc41f5caca908cd1530a3da9131&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.5300 to 1.5340 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR