1-Phenyl-2-thiourea 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Phenylthiourea, 1-Phenylthiourea

20725-0050 20725-0250 20725-1000
TS20725-0050EA 44 USD
TS20725-0050 TS20725-0250 TS20725-1000
1-Phenyl-2-thiourea 97%
1-Phenyl-2-thiourea
Formula: C₇H₈N₂S
Boiling Pt: 295…297 °C (1013 hPa)
Melting Pt: 148…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00004933
CAS Number: 103-85-5
EINECS: 203-151-2
UN: 2811
ADR: 6.1,I
Merck Index: 12,07468

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR