(R)-(−)-Phenylephrine hydrochloride 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
L(-)-Phenylephrine hydrochloride, (R)-(-)-3-[1-Hydroxy-2-(methylamino)ethyl]phenol hydrochloride, (R)-(-)-3-Hydroxy-alpha-(methylaminomethyl)benzyl alcohol hydrochloride, (R)-Phenylephrine hydrochloride, (R)-(-)-1-(3-Hydroxyphenyl)-2-(methylamino)ethanol hydrochloride, R)-(−)-3-Hydroxy-α-(methylaminomethyl)benzyl alcohol hydrochloride
20724-0100
20724-0250
20724-1000
TS20724-0100EA
91
USD
TS20724-0100
TS20724-0250
TS20724-1000
(R)-(−)-Phenylephrine hydrochloride 99%
(R)-(−)-Phenylephrine hydrochloride
Formula:
C₉H₁₃NO₂·HCl Melting Pt: 140…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012605 CAS Number: 61-76-7 |
Specification Test Results
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 140 to 145 °C |
Loss on drying | ≤1 % (1 g, 105 °C) |
Specific optical rotation | -43° to -47° (20 °C, 589 nm) (c=2, H2O) (on dried substance) |
Titration with HClO4 | ≥98.5 % (On dried substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...