L-Adrenaline 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: L-Epinephrine

20440-0250 20440-0010 20440-0050
TS20440-0250EA 396 USD
TS20440-0250 TS20440-0010 TS20440-0050
L-Adrenaline 99%
L-Adrenaline
Formula: C₉H₁₃NO₃
MW: 183.21 g/mol
Melting Pt: 208…211 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002204
CAS Number: 51-43-4
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 208 to 211 °C
Loss on drying ≤1 % (18 hrs) (0.7 kPa, P2O5)
Sulfated ash ≤0.1 %
Specific optical rotation -50° to -53° (20 °C, 589 nm) (c=4, 1 M HCl) (on dried substance)
Titration with HClO4 ≥98.5 % (On dried substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR