Diethylstilbestrol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (E)-3,4-Bis(4-hydroxyphenyl)-3-hexene, Stilbestrol, DES, (E)-4,4'-(Hex-3-ene-3,4-diyl)diphenol

20413-0010 20413-0050 20413-0100
TS20413-0010EA 94 USD
TS20413-0010 TS20413-0050 TS20413-0100
Diethylstilbestrol 99%
Diethylstilbestrol
Formula: C₁₈H₂₀O₂
MW: 268.356 g/mol
Melting Pt: 169…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00002373
CAS Number: 56-53-1

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 169 to 175 °C
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR