2,4-Toluenediisocyanate 75%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Methyl-m-phenylene diisocyanate, Tolylene 2,4-diisocyanate, 2,4-TDI

20250-2500 20250-0010 20250-0025
TS20250-2500EA 59 USD
TS20250-2500 TS20250-0010 TS20250-0025
2,4-Toluenediisocyanate 75%, Technical Grade
2,4-Toluenediisocyanate
Formula: C₉H₆N₂O₂
MW: 174.16 g/mol
Boiling Pt: 115…120 °C (10 mmHg)
Melting Pt: 12.5…13.5 °C
Density: 1.22 g/cm³
Flash Pt: 127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002011
CAS Number: 584-84-9
EINECS: 247-722-4
UN: 2078
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5660 to 1.5700 (20°C, 589 nm)
NMR 10 to 30 % Tolylene 2,6-diisocyanate

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR