(±)-1,2-Octanediol 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Octanediol, (±)-1,2-Octylene glycol, (±)-1,2-Dihydroxy octane

20014-0100 20014-0500
TS20014-0100EA 30 USD
TS20014-0100 TS20014-0500
(±)-1,2-Octanediol 98+%
(±)-1,2-Octanediol
Formula: C₈H₁₈O₂
MW: 146.23 g/mol
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: 28…31 °C
Flash Pt: 138 °C
MDL Number: MFCD00010738
CAS Number: 1117-86-8
EINECS: 214-254-7

Order Now

Specification Test Results

Appearance Colorless to white Low melting solid
Infrared spectrum Conforms
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR