α,α,α-Trifluoro-p-toluic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Trifluoromethyl)benzoic acid

18846-0010 18846-0050
TS18846-0010EA 27 USD
TS18846-0010 TS18846-0050
α,α,α-Trifluoro-p-toluic acid 98%
α,α,α-Trifluoro-p-toluic acid
Formula: C₈H₅F₃O₂
Melting Pt: 219…222 °C
MDL Number: MFCD00002562
CAS Number: 455-24-3
EINECS: 207-242-8

Order Now

Specification Test Results

Appearance White to light grey Crystalline powder
Infrared spectrum Conforms
Melting point 219 to 222 °C
GC ≥97.5 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR