2,6-Difluoroaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

18827-0050 18827-0250 188271000
TS18827-0050EA 42 USD
TS18827-0050 TS18827-0250 TS188271000
2,6-Difluoroaniline 97%
2,6-Difluoroaniline
Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 51…52 °C (15 mmHg)
Density: 1.199 g/cm³
Flash Pt: 43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007655
CAS Number: 5509-65-9
EINECS: 226-853-0
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5070 to 1.5090 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR