3-(Hydroxymethyl)benzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning

18820-0020 18820-0100
TS18820-0020EA 45 USD
TS18820-0020 TS18820-0100
3-(Hydroxymethyl)benzyl alcohol 98%
3-(Hydroxymethyl)benzyl alcohol
Formula: C₈H₁₀O₂
Boiling Pt: 154…159 °C (13 mmHg)
Melting Pt: 56…60 °C
MDL Number: MFCD00004649
CAS Number: 626-18-6
EINECS: 210-934-2

Order Now

Specification Test Results

Appearance White to beige Powder and/or chunks
Infrared spectrum Conforms
Melting point 54 to 59 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR