4-Bromothioanisole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-4-(methylthio)benzene, 4-Bromophenyl methyl sulphide

18819-0050 18819-0250 18819-1000
TS18819-0050EA 26 USD
TS18819-0050 TS18819-0250 TS18819-1000
4-Bromothioanisole 97%
4-Bromothioanisole
Formula: C₇H₇BrS
MW: 203.1 g/mol
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 35…40 °C
Flash Pt: >110 °C
MDL Number: MFCD00000102
CAS Number: 104-95-0
EINECS: 203-255-8
UN: 3335
ADR: 9,III

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals and/or chunks or low melting crystalline mass
Infrared spectrum Conforms
Melting point 35 to 40 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR