α,4-Dichloro-2-nitrotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Chloro-2-nitrobenzyl chloride
18795-0010
18795-0050
TS18795-0010EA
49
USD
TS18795-0010
TS18795-0050
α,4-Dichloro-2-nitrotoluene 98%
α,4-Dichloro-2-nitrotoluene
Formula:
C₇H₅Cl₂NO₂ Melting Pt: 38…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007216 CAS Number: 938-71-6 UN: 3261 ADR: 8,III |
Specification Test Results
Appearance | Light yellow to tan Low melting solid |
Infrared spectrum | Conforms |
Melting point | 39 to 44 °C |
GC | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...