3-Phenoxypropan-1-ol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Phenoxy-1-propanol

18698-0050 18698-0250
TS18698-0050EA 49 USD
TS18698-0050 TS18698-0250
3-Phenoxypropan-1-ol 97%
3-Phenoxypropan-1-ol
Formula: C₉H₁₂O₂
MW: 152.193 g/mol
Boiling Pt: 83…85 °C
Density: 1.04 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00039549
CAS Number: 6180-61-6

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5280 to 1.5300 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR