m-Phenetidine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Amino-3-ethoxybenzene, 3-Ethoxyaniline
18672-0050
18672-0250
TS18672-0050EA
50
USD
TS18672-0050
TS18672-0250
m-Phenetidine 98%
m-Phenetidine
Formula:
C₈H₁₁NO Boiling Pt: 248 °C (1013 hPa) Density: 1.03 g/cm³ Flash Pt: >112 °C Storage Temperature: Ambient |
CAS Number:
621-33-0 EINECS: 210-680-2 UN: 2311 ADR: 6.1,III |
Specification Test Results
Appearance | Clear brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5650 to 1.5670 (20°C, 589 nm) |
GC | ≥97.5 % |
HPLC | ≥97.5 % |
Residue after ignition | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...