3,4-Dimethoxyphenol 97%

Supplier: Thermo Scientific Chemicals
Warning

18670-0100 18670-0500
TS18670-0100EA 172 USD
TS18670-0100 TS18670-0500
3,4-Dimethoxyphenol 97%
3,4-Dimethoxyphenol
Formula: C₈H₁₀O₃
Melting Pt: 77…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00008390
CAS Number: 2033-89-8
EINECS: 217-995-4

Order Now

Specification Test Results

Appearance Off-white to light beige Crystalline powder (May darken on storage)
Infrared spectrum Conforms
Melting point 77 to 82 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR