Dimethyl isophthalate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Isophthalic acid dimethyl ester

18651-0025 18651-0250 18651-5000
TS18651-0025EA 160 USD
TS18651-0025 TS18651-0250 TS18651-5000
Dimethyl isophthalate 99%
Dimethyl isophthalate
Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Boiling Pt: 124 °C (16hPa)
Melting Pt: >66 °C
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00008433
CAS Number: 1459-93-4
EINECS: 215-951-9

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 60 to 70 °C
GC ≥98.5 %
Solubility (2 % in DMSO) Clear solution
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR