2-(2-Chlorophenyl)ethanol 98%

Supplier: Thermo Scientific Chemicals

18619-0010 18619-0050 18619-0250
TS18619-0010EA 27 USD
TS18619-0010 TS18619-0050 TS18619-0250
2-(2-Chlorophenyl)ethanol 98%
2-(2-Chlorophenyl)ethanol
Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 84…85 °C (3 mmHg)
Density: 1.19 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002888
CAS Number: 19819-95-5
EINECS: 243-348-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5495 to 1.5515 (20°C, 589 nm)
GC ≥97.5 %
Residue after ignition ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR