(±)-1-Indanol 98%

Supplier: Thermo Scientific Chemicals
Warning

18611-0100 18611-0500
TS18611-0100EA 103 USD
TS18611-0100 TS18611-0500
(±)-1-Indanol 98%
(±)-1-Indanol
Formula: C₉H₁₀O
Melting Pt: 50…54 °C
Flash Pt: 145 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003797
CAS Number: 6351-10-6
EINECS: 228-755-3
UN: 2811

Order Now

Specification Test Results

Appearance White to yellow to beige to light brown Powder or crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 50 to 55 °C
GC ≥97.5 %
HPLC ≥97.5 %
Specific optical rotation +7° to +10° (20 °C, 589 nm) (1% in MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR