1,4-Diiodobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

18597-0500 18597-2500
TS18597-0500EA 111 USD
TS18597-0500 TS18597-2500
1,4-Diiodobenzene 98%
1,4-Diiodobenzene
Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 285 °C (760 mmHg)
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00001054
CAS Number: 624-38-4
EINECS: 210-842-2

Order Now

Specification Test Results

Appearance White to beige to tan Powder or crystals
Infrared spectrum Conforms
Melting point 128 to 133 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR