α,α,α,α',α',α'-Hexafluoro-3,5-xylidine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Di(trifluoromethyl)aniline, 3,5-Bis(trifluoromethyl)aniline

18569-0100 18569-0500
TS18569-0100EA 44 USD
TS18569-0100 TS18569-0500
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine 98+%
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine
Formula: C₈H₅F₆N
MW: 229.12 g/mol
Boiling Pt: 85 °C (15 mmHg)
Melting Pt: 3 °C
Density: 1.47 g/cm³
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00000394
CAS Number: 328-74-5
EINECS: 206-335-0

Order Now

Specification Test Results

Appearance Clear light yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4350 (20°C, 589 nm)
Titration with HClO4 ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR