4-Bromo-2,6-xylidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-2,6-dimethylaniline

18519-0050 18519-0250
TS18519-0050EA 27 USD
TS18519-0050 TS18519-0250
4-Bromo-2,6-xylidine 98%
4-Bromo-2,6-xylidine
Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 120 °C (0.2 mmHg)
Melting Pt: 48…52 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007826
CAS Number: 24596-19-8
EINECS: 246-337-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Beige to brown to purple Powder or crystals
Infrared spectrum Conforms
Melting point 48 to 52 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR