3-Nitrobenzylamine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Nitrobenzylammonium hydrochloride
18473-0010
18473-0050
TS18473-0010EA
53
USD
TS18473-0010
TS18473-0050
3-Nitrobenzylamine hydrochloride 97%
3-Nitrobenzylamine hydrochloride
Formula:
C₇H₉ClN₂O₂ MW: 188.62 g/mol Melting Pt: 226…231 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012858 CAS Number: 26177-43-5 EINECS: 247-502-8 |
Specification Test Results
Appearance | Off-white to tan Solid |
Infrared spectrum | Conforms |
Melting point | 226 to 231 °C |
Titration with HClO4 | ≥96.0 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...