4-Ethylbenzoic acid 97%
Supplier: Thermo Scientific Chemicals
18449-0050
18449-0250
TS18449-0050EA
14
USD
TS18449-0050
TS18449-0250
4-Ethylbenzoic acid 97%
4-Ethylbenzoic acid
Formula:
C₉H₁₀O₂ |
MDL Number:
MFCD00002570 CAS Number: 619-64-7 |
Specification Test Results
Appearance | White to beige Powder |
Infrared spectrum | Conforms |
Melting point | 110 to 114 °C |
Titration with TBAH | ≥96.0 % |
pH | 11 to 12 (1 % in water) |
Specific optical rotation | -9° to -13° (20 °C, 589 nm) (4 % in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...