O-Dianisidine dihydrochloride 98%

Supplier: Thermo Scientific Chemicals
Danger

18447-0050 18447-0250
TS18447-0050EA 56 USD
TS18447-0050 TS18447-0250
O-Dianisidine dihydrochloride 98%
O-Dianisidine dihydrochloride
Formula: C₁₄H₁₈Cl₂N₂O₂
MW: 317.21 g/mol
Melting Pt: 268 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012488
CAS Number: 20325-40-0
EINECS: 243-737-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to beige to grey to purple-grey Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
Titration with HClO4 ≥97.5 % (on dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR