(2-Chlorophenyl)acetic acid 98.5+%

Supplier: Thermo Scientific Chemicals
Warning

18403-0050 18403-0250 18403-1000
TS18403-0050EA 21 USD
TS18403-0050 TS18403-0250 TS18403-1000
(2-Chlorophenyl)acetic acid 98.5+%
(2-Chlorophenyl)acetic acid
Formula: (C₆H₄Cl)CH₂COOH
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00004317
CAS Number: 2444-36-2
EINECS: 219-482-0

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 94 to 97 °C
HPLC ≥98.50 %
Titration with TBAH ≥98.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR