3-Phenoxybenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(Hydroxymethyl)diphenyl ether, (3-Phenoxyphenyl)methanol

18377-0050 18377-0250 18377-1000
TS18377-0050EA 26 USD
TS18377-0050 TS18377-0250 TS18377-1000
3-Phenoxybenzyl alcohol 98%
3-Phenoxybenzyl alcohol
Formula: C₁₃H₁₂O₂
MW: 200.24 g/mol
Boiling Pt: 135…140 °C (1 mmHg)
Density: 1.1456 g/cm³
Flash Pt: 309 °C
MDL Number: MFCD00004636
CAS Number: 13826-35-2
EINECS: 237-525-1

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5910 to 1.5950 (20°C, 589 nm)
GC ≥97.5 %
Solubility (2 % in DMSO) Clear colorless to light yellow solution
Titration with HClO4 99 to 101 % (quetiapine hemifumarate dry substance)
Titration with NaOH 12.5 to 13.8 % (fumaric acid dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR