1-Phenyl-2-propanol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (2-Hydroxypropyl)benzol, (±)-1-Phenyl-2-propanol, (±)-α-Methylphenethyl alcohol, Benzylmethylcarbinol

18306-0050 18306-0250
TS18306-0050EA 41 USD
TS18306-0050 TS18306-0250
1-Phenyl-2-propanol 99%
1-Phenyl-2-propanol
Formula: C₆H₅CH₂CH(OH)CH₃
MW: 136.19 g/mol
Density: 0.99 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00004540
CAS Number: 698-87-3
EINECS: 211-821-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5200 to 1.5220 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR