2-(3-Methoxyphenyl)ethanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methoxyphenethyl alcohol

18225-0010 18225-0050 182250250
TS18225-0010EA 68 USD
TS18225-0010 TS18225-0050 TS182250250
2-(3-Methoxyphenyl)ethanol 97%
2-(3-Methoxyphenyl)ethanol
Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 141…143 °C (12 mmHg)
Density: 1.075 g/cm³
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002893
CAS Number: 5020-41-7

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure
Titration with HClO4 97.5 to 102.5 % (dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR