2,5-Dimethoxybenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals

18220-0050 18220-0250
TS18220-0050EA 63 USD
TS18220-0050 TS18220-0250
2,5-Dimethoxybenzyl alcohol 99%
2,5-Dimethoxybenzyl alcohol
Formula: C₉H₁₂O₃
Boiling Pt: 122…125 °C (1 mmHg)
Flash Pt: >110 °C
MDL Number: MFCD00004615
CAS Number: 33524-31-1
EINECS: 251-562-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5460 to 1.5480 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation +68° to +78° (20°C, 589 nm) (1% in CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR