2,6-Dichlorobenzoyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,6-Dichlorobenzoyl Chloride

18206-0100 18206-0500
TS18206-0100EA 50 USD
TS18206-0100 TS18206-0500
2,6-Dichlorobenzoyl chloride 99%
2,6-Dichlorobenzoyl chloride
Formula: C₇H₃Cl₃O
MW: 209.46 g/mol
Boiling Pt: 142…143 °C (21 mmHg)
Melting Pt: 18 °C
Density: 1.45 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000662
CAS Number: 4659-45-4
EINECS: 225-102-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow to light pink Liquid
Infrared spectrum Conforms
Refractive index 1.5590 to 1.5610 (20°C, 589 nm)
GC ≥98.5 %
Titration RCOCl ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR