9-Anthracenemethanol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 9-Anthrylmethanol, Anthracene-9-methanol

18187-0050 18187-0250
TS18187-0050EA 56 USD
TS18187-0050 TS18187-0250
9-Anthracenemethanol 98%
9-Anthracenemethanol
Formula: C₁₅H₁₂O
MW: 208.26 g/mol
Boiling Pt: 371
Melting Pt: 160…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00001264
CAS Number: 1468-95-7
EINECS: 215-998-5

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 160 to 164 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR