Cinnamyl chloride 95% (trans-isomers)
Supplier: Thermo Scientific Chemicals
|
Danger
|
18185-0050
18185-1000
18185-5000
TS18185-0050EA
23
USD
TS18185-0050
TS18185-1000
TS18185-5000
Cinnamyl chloride 95% (trans-isomers)
Cinnamyl chloride
Formula:
C₉H₉Cl Boiling Pt: 120…126 °C (15 mmHg) Melting Pt: 7 °C Density: 1.096 g/cm³ Flash Pt: 79 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000986 CAS Number: 2687-12-9 UN: 2922 ADR: 8,II |
Specification Test Results
Appearance | Colorless to yellow to yellow-brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5830 to 1.5850 (20°C, 589 nm) |
GC | ≥94 % |
HPLC | ≥97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...