α,2-Dibromotoluene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Bromobenzyl bromide

18179-0250 18179-1000
TS18179-0250EA 87 USD
TS18179-0250 TS18179-1000
α,2-Dibromotoluene 98%
α,2-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 129 °C (19 mmHg)
Melting Pt: 29…32 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000173
CAS Number: 3433-80-5
EINECS: 222-334-8
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.6170 to 1.6190 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR