α,3-Dibromotoluene 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Bromobenzyl bromide, 1-Bromo-3-(bromomethyl)benzene

18178-0050 18178-0250
TS18178-0050EA 39 USD
TS18178-0050 TS18178-0250
α,3-Dibromotoluene 95%
α,3-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 130 °C (12 mmHg)
Melting Pt: 37…42 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000176
CAS Number: 823-78-9
EINECS: 212-519-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White Adhering crystalline powder or crystals
Infrared spectrum Conforms
Melting point 37 to 42 °C
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR