4-Bromobenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Bromophenyl)methanol

18177-0100 18177-0500
TS18177-0100EA 61 USD
TS18177-0100 TS18177-0500
4-Bromobenzyl alcohol 99%
4-Bromobenzyl alcohol
Formula: C₇H₇BrO
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 75…78 °C
MDL Number: MFCD00004650
CAS Number: 873-75-6
EINECS: 212-851-7

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 75 to 78 °C
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR