4-Chlorophenol 99+%

Supplier: Thermo Scientific Chemicals
Warning

18100-0025 18100-0051 18100-1000 18100-5000
TS18100-0025EA 321 USD
TS18100-0025 TS18100-0051 TS18100-1000 TS18100-5000
4-Chlorophenol 99+%
4-Chlorophenol
Formula: C₆H₅ClO
Boiling Pt: 220 °C (760 mmHg)
Melting Pt: 41…45 °C
Density: 1.26 g/cm³
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00002318
CAS Number: 106-48-9
EINECS: 203-402-6
UN: 2020
ADR: 6.1,III
Merck Index: 13,02173

Order Now

Specification Test Results

Appearance White to light brown Crystals or fused mass
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR