1-Chloronaphthalene 85% remainder 2-Chloronaphthalene, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Triisodecyl phosphite, α-Chloronaphthalene
18097-0010
18097-0025
18097-1000
TS18097-0010EA
318
USD
TS18097-0010
TS18097-0025
TS18097-1000
1-Chloronaphthalene 85% remainder 2-Chloronaphthalene, Technical Grade
1-Chloronaphthalene
Formula:
C₁₀H₇Cl Boiling Pt: 259 °C (760 mmHg) Melting Pt: –2.5 °C Density: 1.194 g/cm³ Flash Pt: 121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003874 CAS Number: 90-13-1 EINECS: 201-967-3 UN: 3082 ADR: 9,III Merck Index: 13,02168 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.6300 to 1.6340 (20°C, 589 nm) |
HPLC | ≥93 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...