Phenol 99+%, loose crystals for biochemistry
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Phenylalcohol, Carbolic acid, Hydroxybenzene
18078-0015
18078-1000
18078-5000
TS18078-0015EA
480
USD
TS18078-0015
TS18078-1000
TS18078-5000
Phenol 99+%, loose crystals for biochemistry
Phenol
Formula:
C₆H₅OH MW: 94.11 g/mol Boiling Pt: 181.7 Melting Pt: 40.8 Density: 1.07 g/cm³ Flash Pt: 79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002143 CAS Number: 108-95-2 UN: 1671 ADR: 6.1 II Merck Index: 12,07390 |
Specification Test Results
Appearance | Colorless to white Loose crystals or flakes |
Infrared spectrum | Conforms |
Water | ≤0.5 % |
GC | ≥99.0 % |
Residue after evaporation | ≤0.05 % |
Solidification point | ≥40.5°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...