Dimethyl terephthalate 99%
Supplier: Thermo Scientific Chemicals
18058-0010
18058-0025
18058-0250
18058-5000
TS18058-0010EA
48
USD
TS18058-0010
TS18058-0025
TS18058-0250
TS18058-5000
Dimethyl terephthalate 99%
Dimethyl terephthalate
Formula:
C₁₀H₁₀O₄ Boiling Pt: 288 °C (1013 hPa) Melting Pt: 140…143 °C Flash Pt: 154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008440 CAS Number: 120-61-6 EINECS: 204-411-8 Merck Index: 12,09306 |
Specification Test Results
Appearance | White Flakes and/or pellets |
Infrared spectrum | Conforms |
Melting point | 140 to 143 °C |
GC | ≥98.5 % |
Acid value | ≤0.03 mg KOH/g |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...