4-Chlorosalicylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloro-2-hydroxybenzoic acid

17984-0050 17984-0250 17984-1000
TS17984-0050EA 27 USD
TS17984-0050 TS17984-0250 TS17984-1000
4-Chlorosalicylic acid 98%
4-Chlorosalicylic acid
Formula: C₇H₅ClO₃
MW: 172.57 g/mol
Melting Pt: 209…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00002449
CAS Number: 5106-98-9
EINECS: 225-829-7

Order Now

Specification Test Results

Appearance Off-white to light beige Powder
Infrared spectrum Conforms
Melting point 209 to 213 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR